Bromothymol blue ACS

Supplier: BeanTown Chemical

Synonyms: 3′,3″-Dibromothymolsulphonphthalein, 3,3-Bis(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

143485-25G 143485-5G
BT143485-25GEA 98.95 USD
BT143485-25G BT143485-5G
Bromothymol blue ACS
Bromothymol blue

CAS: 76-59-5; EC No: 200-971-2; MDL No: MFCD00005872; RTECS: SJ4500000 Crystalline; Molecular Formula: C27H28Br2O5S; MW: 624.38 Melting Point: 200-202°


  • RTECS: SJ4500000
  • Crystalline

Formula: C₂₇H₂₈Br₂O₅S
MW: 624.39 g/mol
Melting Pt: 200…202 °C
MDL Number: MFCD00005872
CAS Number: 76-59-5
EINECS: 200-971-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR