1-Bromohexadecane ≥98%

Supplier: BeanTown Chemical

143415-1KG 143415-250G
BT143415-1KGEA 118.98 USD
BT143415-1KG BT143415-250G
1-Bromohexadecane ≥98%
1-Bromohexadecane

CAS: 112-82-3; EC No: 204-008-7; MDL No: MFCD00000230 Liquid/Low Melting Solid; Linear Formula: CH3(CH2)15Br ; Molecular Formula: C16H33Br; MW: 305.34 Melting Point: 16-18°; Boiling Point: 190°/11 mmHg; Flash point: 177°C (351°F) Density (g/mL): 0.999; Refractive Index: 1.461


  • Refractive index: 1.461
  • Liquid/Low Melting Solid

Formula: BrH₂C(CH₂)₁₄CH₃
MW: 305.34 g/mol
Boiling Pt: 190 °C/11 mmHg
Melting Pt: 16…18 °C
Density: 0.999 g/mL
Flash Pt: 177 °C (351 °F)
MDL Number: MFCD00000230
CAS Number: 112-82-3
EINECS: 204-008-7

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR