Manganese(II) oxide ≥99%

Supplier: BeanTown Chemical

143380-2KG 143380-500G
BT143380-2KGEA 154 USD
BT143380-2KG BT143380-500G
Manganese(II) oxide ≥99%
Manganese(II) oxide

CAS: 1344-43-0; EC No: 215-695-8; MDL No: MFCD00016226; RTECS: OP0900000 +200 Mesh Powder; Molecular Formula: MnO; MW: 70.94 Melting Point: 450° Density (g/mL): 5.45


  • RTECS: OP0900000
  • 200 Mesh Powder

Formula: MnO
Melting Pt: >450 °C
Density: 5.45 g/mL
MDL Number: MFCD00016226
CAS Number: 1344-43-0
EINECS: 215-695-8

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR