Cerium(III) acetylacetonate hydrate

Supplier: BeanTown Chemical

Synonyms: Cerium(III) 2,4-pentanedionate hydrate

143350-100G 143350-500G
BT143350-100GEA 175 USD
BT143350-100G BT143350-500G
Cerium(III) acetylacetonate hydrate
Cerium(III) acetylacetonate hydrate

CAS: 206996-61-4; EC No: 239-720-7; MDL No: MFCD00150164 Powder; Molecular Formula: C15H21CeO6·xH2O; MW: 437.44 (anhydrous) Hygroscopic


  • Hygroscopic
  • Powder

Formula: C₁₅H₂₁CeO₆·xH₂O
MW: 437.44 g/mol (anhy)
MDL Number: MFCD00150164
CAS Number: 206996-61-4
EINECS: 239-720-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR