3-Cyclopropyl-1H-pyrazol-5-amine ≥96%

Supplier: BeanTown Chemical

Synonyms: (5-Amino-1H-pyrazol-3-yl)cyclopropane, 5-Amino-3-cyclopropyl-1H-pyrazole, 3-Cyclopropyl-1H-pyrazol-5-amine

143340-1G 143340-250MG
BT143340-1GEA 260 USD
BT143340-1G BT143340-250MG
3-Cyclopropyl-1H-pyrazol-5-amine ≥96%
3-Cyclopropyl-1H-pyrazol-5-amine

CAS: 175137-46-9; MDL No: MFCD08461681 Liquid; Molecular Formula: C6H9N3; MW: 123.16 Air Sensitive


  • Air Sensitive
  • Liquid

Formula: C₆H₉N₃
MW: 123.159 g/mol
MDL Number: MFCD08461681
CAS Number: 175137-46-9

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR