N-(4-Oxocyclohexyl)phthalimide ≥98%

Supplier: BeanTown Chemical

Synonyms: 4-Phthalimidocyclohexanone, 2-(4-Oxocyclohexyl)isoindoline-1,3-dione

143325-25G 143325-5G
BT143325-25GEA 233 USD
BT143325-25G BT143325-5G
N-(4-Oxocyclohexyl)phthalimide ≥98%
N-(4-Oxocyclohexyl)phthalimide

CAS: 104618-32-8 Solid; Molecular Formula: C14H13NO3; MW: 243.26 Density (g/mL): 1.357


  • Solid

Formula: C₁₄H₁₃NO₃
Density: 1.357 g/mL
MDL Number: MFCD00158659
CAS Number: 104618-32-8

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR