Aluminum silicate
Supplier: BeanTown Chemical
143270-2KG
143270-500G
BT143270-2KGEA
92
USD
BT143270-2KG
BT143270-500G
Aluminum silicate
Aluminum silicate
CAS: 12428-46-5; MDL No: MFCD00062311
Powder; Linear Formula: Al2Si2O7·2H2O; MW: 258.16
- Powder
Formula:
Al₂Si₂O₇·2H₂O |
MDL Number:
MFCD00062311 CAS Number: 12428-46-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...