Lanthanum(III) oxide ≥99.9% (trace rare earth metals basis)

Supplier: BeanTown Chemical

143265-100G 143265-500G
BT143265-100GEA 85.57 USD
BT143265-100G BT143265-500G
Lanthanum(III) oxide ≥99.9% (trace rare earth metals basis)
Lanthanum(III) oxide

CAS: 1312-81-8; EC No: 215-200-5; MDL No: MFCD00011071 Powder; Molecular Formula: La2O3; MW: 325.82 Melting Point: 2307° Density (g/mL): 6.51 Hygroscopic


  • Hygroscopic
  • Powder

Formula: La₂O₃
MW: 325.81 g/mol
Melting Pt: 2307 °C
Density: 6.51 g/mL
MDL Number: MFCD00011071
CAS Number: 1312-81-8
EINECS: 215-200-5

Order Now

Specification Test Results

Assay 99.9%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR