Calcium hexaboride ≥99% (trace metals basis)
Supplier: BeanTown Chemical
143255-100G
143255-25G
BT143255-100GEA
261
USD
BT143255-100G
BT143255-25G
Calcium hexaboride ≥99% (trace metals basis)
Calcium hexaboride
CAS: 12007-99-7; EC No: 234-525-3; MDL No: MFCD00148904
-20 Mesh Powder; Linear Formula: CaB6; MW: 104.94
Density (g/mL): 2.3
- -20 Mesh Powder
Formula:
B₆Ca MW: 104.94 g/mol Density: 2.3 g/mL |
MDL Number:
MFCD00148904 CAS Number: 12007-99-7 EINECS: 234-525-3 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...