2,3-Dihydroxynaphthalene ≥98%
Supplier: BeanTown Chemical
Synonyms:
Naphthalene-2,3-diol, 2,3-Naphthalenediol
143240-250G
143240-50G
BT143240-250GEA
169
USD
BT143240-250G
BT143240-50G
2,3-Dihydroxynaphthalene ≥98%
2,3-Dihydroxynaphthalene
CAS: 92-44-4; EC No: 202-156-7; MDL No: MFCD00004073; RTECS: QJ4750000
Crystalline; Molecular Formula: C10H8O2; MW: 160.17
Melting Point: 161-165°
- RTECS: QJ4750000
- Crystalline
Formula:
C₁₀H₈O₂ MW: 160.17 g/mol Melting Pt: 161…165 °C |
MDL Number:
MFCD00004073 CAS Number: 92-44-4 EINECS: 202-156-7 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...