Yttrium(III) trifluoroacetate hydrate ≥99.9% (trace rare earth metals basis)
Supplier: BeanTown Chemical
143230-10G
143230-50G
BT143230-10GEA
69
USD
BT143230-10G
BT143230-50G
Yttrium(III) trifluoroacetate hydrate ≥99.9% (trace rare earth metals basis)
Yttrium(III) trifluoroacetate hydrate
CAS: 304851-95-4; MDL No: MFCD05663780
Crystalline; Linear Formula: Y(CF3COO)3·xH2O; Molecular Formula: C6F9O6Y·xH2O; MW: 427.95 (anhydrous)
- Crystalline
Formula:
C₆F₉O₆Y·xH₂O MW: 427.951 g/mol (anhy) |
MDL Number:
MFCD05663780 CAS Number: 304851-95-4 |
Specification Test Results
Assay | 99.9% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...