Iron(III) acetylacetonate

Supplier: BeanTown Chemical

Synonyms: Tris(2,4-pentanedionato-O,O')iron (III), Ferric acetylacetonate, Iron (III) 2,4-pentanedionate, Iron (III) tris(2,4-pentanedionato-O,O')

143220-100G 143220-500G
BT143220-100GEA 65.12 USD
BT143220-100G BT143220-500G
Iron(III) acetylacetonate
Iron(III) acetylacetonate

CAS: 14024-18-1; EC No: 237-853-5; MDL No: MFCD00000020; RTECS: NO8960000 Powder; Molecular Formula: C15H21FeO6; MW: 353.17 Melting Point: 180-182° (decomposes) Density (g/mL): 5.24


  • RTECS: NO8960000
  • Powder

Formula: C₁₅H₂₁FeO₆
MW: 353.18 g/mol
Boiling Pt: 100
Melting Pt: 180…182 °C (dec.)
Density: 5.24 g/mL
MDL Number: MFCD00000020
CAS Number: 14024-18-1
EINECS: 237-853-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR