(4-Nitrophenyl)-β-D-glucopyranoside ≥98%
Supplier: BeanTown Chemical
143210-1G
143210-5G
BT143210-1GEA
64.29
USD
BT143210-1G
BT143210-5G
(4-Nitrophenyl)-β-D-glucopyranoside ≥98%
(4-Nitrophenyl)-β-D-glucopyranoside
CAS: 2492-87-7; EC No: 219-661-3; MDL No: MFCD00006593
Crystalline; Molecular Formula: C12H15NO8; MW: 301.25
- Crystalline
Formula:
C₁₂H₁₅NO₈ MW: 301.25 g/mol |
MDL Number:
MFCD00006593 CAS Number: 2492-87-7 EINECS: 219-661-3 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...