Tantalum(V) oxide ≥99.5% (trace metals basis)
Supplier: BeanTown Chemical
143195-100G
143195-25G
BT143195-100GEA
312.37
USD
BT143195-100G
BT143195-25G
Tantalum(V) oxide ≥99.5% (trace metals basis)
Tantalum(V) oxide
CAS: 1314-61-0; EC No: 215-238-2; MDL No: MFCD00011254
Powder; Linear Formula: Ta2O5; MW: 441.89
Melting Point: 1800°
Density (g/mL): 8.2
- Powder
Formula:
O₅Ta₂ Melting Pt: 1800 °C Density: 8.2 g/mL |
MDL Number:
MFCD00011254 CAS Number: 1314-61-0 EINECS: 215-238-2 |
Specification Test Results
Assay | 99.5% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...