(2-Chloro-4-pyridinyl)methanol ≥97%

Supplier: BeanTown Chemical

Synonyms: (2-Chloropyridin-4-yl)methanol, 2-Chloro-4-(hydroxymethyl)pyridine, 2-Chloropyridine-4-methanol

143175-1G 143175-5G
BT143175-1GEA 205 USD
BT143175-1G BT143175-5G
(2-Chloro-4-pyridinyl)methanol ≥97%
(2-Chloro-4-pyridinyl)methanol

CAS: 100704-10-7; MDL No: MFCD06858767 Solid; Molecular Formula: C6H6ClNO; MW: 143.57 Melting Point: 59-64°


  • Solid

Formula: C₆H₆ClNO
MW: 143.57 g/mol
Melting Pt: 59…64 °C
MDL Number: MFCD06858767
CAS Number: 100704-10-7

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR