1-Ethyl-4-[(4-methoxyphenyl)ethynyl]benzene ≥99%
Supplier: BeanTown Chemical
Synonyms:
1-(4-Ethylphenyl)-2-(4-methoxyphenyl)acetylene, 4-[(4-Ethylphenyl)ethynyl]anisole, 1-Ethyl-4-((4-methoxyphenyl)ethynyl)benzene
143155-1G
143155-5G
BT143155-1GEA
58
USD
BT143155-1G
BT143155-5G
1-Ethyl-4-[(4-methoxyphenyl)ethynyl]benzene ≥99%
1-Ethyl-4-[(4-methoxyphenyl)ethynyl]benzene
CAS: 63221-88-5; MDL No: MFCD00807428
Crystalline/Powder; Molecular Formula: C17H16O; MW: 236.31
Melting Point: 35°; Boiling Point: 151-153°/1 mmHg
- Crystalline/Powder
Formula:
C₁₇H₁₆O MW: 236.31 g/mol Boiling Pt: 151…153 °C/1 mmHg Melting Pt: 35 °C |
MDL Number:
MFCD00807428 CAS Number: 63221-88-5 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...