5-Aminolevulinic acid hydrochloride ≥99%

Supplier: BeanTown Chemical

Synonyms: 5-Amino-4-ketovaleric acid hydrochloride, 5-Amino-4-oxopentanoic acid hydrochloride, δ-Aminolevulinic Acid Hydrochloride

143115-100MG 143115-2G 143115-500MG
BT143115-100MGEA 32 USD
BT143115-100MG BT143115-2G BT143115-500MG
5-Aminolevulinic acid hydrochloride ≥99%
5-Aminolevulinic acid hydrochloride

CAS: 5451-09-2; EC No: 226-679-5; MDL No: MFCD00012869; RTECS: OI1640000 Crystalline; Molecular Formula: C5H9NO3·HCl; MW: 167.59 Melting Point: 150° (decomposes) Hygroscopic


  • RTECS: OI1640000
  • Hygroscopic
  • Crystalline

Formula: C₅H₁₀ClNO₃
MW: 167.59 g/mol
Melting Pt: 150 °C (dec.)
MDL Number: MFCD00012869
CAS Number: 5451-09-2
EINECS: 226-679-5

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR