2,2-Difluoro-1,3-benzodioxole-5-carboxylic acid ≥97%
Supplier: BeanTown Chemical
Synonyms:
2,2-Difluorobenzo[d][1,3]dioxole-5-carboxylic acid
143100-1G
143100-5G
BT143100-1GEA
67
USD
BT143100-1G
BT143100-5G
2,2-Difluoro-1,3-benzodioxole-5-carboxylic acid ≥97%
2,2-Difluoro-1,3-benzodioxole-5-carboxylic acid
CAS: 656-46-2; MDL No: MFCD00792417
Liquid; Molecular Formula: C8H4F2O4; MW: 202.12
- Liquid
Formula:
C₈H₄F₂O₄ MW: 202.12 g/mol |
MDL Number:
MFCD00792417 CAS Number: 656-46-2 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...