5-Methylpyridin-3-ol ≥97%

Supplier: BeanTown Chemical

Synonyms: 3-Hydroxy-5-methylpyridine, 3-Hydroxy-5-picoline

143095-1G 143095-250MG 143095-5G
BT143095-1GEA 338 USD
BT143095-1G BT143095-250MG BT143095-5G
5-Methylpyridin-3-ol ≥97%
5-Methylpyridin-3-ol

CAS: 42732-49-0; MDL No: MFCD00661297 Crystalline; Molecular Formula: C6H7NO; MW: 109.13 Melting Point: 136-139°


  • Crystalline

Formula: C₆H₇NO
MW: 109.13 g/mol
Melting Pt: 136…139 °C
MDL Number: MFCD00661297
CAS Number: 42732-49-0

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR