1H,1H,2H,2H-Perfluoro-1-iodohexane ≥97%
Supplier: BeanTown Chemical
143075-25G
143075-5G
BT143075-25GEA
212.87
USD
BT143075-25G
BT143075-5G
1H,1H,2H,2H-Perfluoro-1-iodohexane ≥97%
1H,1H,2H,2H-Perfluoro-1-iodohexane
CAS: 2043-55-2; EC No: 218-055-6; MDL No: MFCD00039409
Liquid; Linear Formula: CF3(CF2)3CH2CH2I; Molecular Formula: C6H4F9I; MW: 373.99
Density (g/mL): 1.94; Refractive Index: 1.370
Light Sensitive
- Refractive index: 1.370
- Light Sensitive
- Liquid
Formula:
C₆H₄F₉I MW: 373.99 g/mol Density: 1.94 g/mL |
MDL Number:
MFCD00039409 CAS Number: 2043-55-2 EINECS: 218-055-6 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...