1-Bromo-3-nitrobenzene ≥97%

Supplier: BeanTown Chemical

143070-25G 143070-5G
BT143070-25GEA 50 USD
BT143070-25G BT143070-5G
1-Bromo-3-nitrobenzene ≥97%
1-Bromo-3-nitrobenzene

CAS: 585-79-5; EC No: 209-563-9; MDL No: MFCD00024298 Crystalline; Molecular Formula: C6H4BrNO2; MW: 202.01 Melting Point: 52-55°; Boiling Point: 256°; Flash point: 110°C (235°F) Density (g/mL): 1.704


  • Crystalline

Formula: C₆H₄BrNO₂
MW: 202.01
Boiling Pt: 256
Melting Pt: 52…55 °C
Density: 1.704 g/mL
Flash Pt: 110 °C (235 °F)
MDL Number: MFCD00024298
CAS Number: 585-79-5
EINECS: 209-563-9
UN: 3459
ADR: 6.1 III

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR