2-Nitrobenzenesulfonamide ≥98%

Supplier: BeanTown Chemical

143015-25G 143015-5G
BT143015-25GEA 194 USD
BT143015-25G BT143015-5G
2-Nitrobenzenesulfonamide ≥98%
2-Nitrobenzenesulphonamide

CAS: 5455-59-4; MDL No: MFCD00009807 Powder; Molecular Formula: C6H6N2O4S; MW: 202.19 Melting Point: 190-192°


  • Powder

Formula: C₆H₆N₂O₄S
MW: 202.19 g/mol
Melting Pt: 190…192 °C
MDL Number: MFCD00009807
CAS Number: 5455-59-4

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR