2-Amino-4-chloropyrimidine ≥98%

Supplier: BeanTown Chemical

Synonyms: 4-chloropyrimidin-2-amine

143010-1G 143010-250MG 143010-5G
BT143010-1GEA 77 USD
BT143010-1G BT143010-250MG BT143010-5G
2-Amino-4-chloropyrimidine ≥98%
2-Amino-4-chloropyrimidine

CAS: 3993-78-0; MDL No: MFCD00038021 Solid; Molecular Formula: C4H4ClN3; MW: 129.55 Melting Point: 155-160° (decomposes)


  • Solid

Formula: C₄H₄ClN₃
MW: 129.55
Melting Pt: 155…160 °C (dec.)
MDL Number: MFCD00038021
CAS Number: 3993-78-0

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR