2,6-Dibromo-3-pyridinecarboxylic acid ≥98%
Supplier: BeanTown Chemical
Synonyms:
2,6-Dibromopyridine-3-carboxylic acid
143005-1G
143005-5G
BT143005-1GEA
86
USD
BT143005-1G
BT143005-5G
2,6-Dibromo-3-pyridinecarboxylic acid ≥98%
2,6-Dibromo-3-pyridinecarboxylic acid
CAS: 55304-85-3; MDL No: MFCD11846496
Solid; Molecular Formula: C6H3NO2Br2; MW: 280.90
Boiling Point: 380°
Density (g/mL): 2.202
- Solid
Formula:
C₆H₃Br₂NO₂ MW: 280.90 g/mol Boiling Pt: 380 °C Density: 2.202 g/mL |
MDL Number:
MFCD11846496 CAS Number: 55304-85-3 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...