(Bromomethyl)cyclohexane ≥98%
Supplier: BeanTown Chemical
142990-25G
142990-5G
BT142990-25GEA
61
USD
BT142990-25G
BT142990-5G
(Bromomethyl)cyclohexane ≥98%
(Bromomethyl)cyclohexane
CAS: 2550-36-9; MDL No: MFCD00001509
Liquid; Molecular Formula: C7H13Br; MW: 177.08
Boiling Point: 76-77°/26 mmHg; Flash point: 57°C (135°F)
Density (g/mL): 1.269; Refractive Index: 1.492
- Refractive index: 1.492
- Liquid
Formula:
C₇H₁₃Br Boiling Pt: 76…77 °C/26 mmHg Density: 1.269 g/mL Flash Pt: 57 °C (135 °F) |
MDL Number:
MFCD00001509 CAS Number: 2550-36-9 UN: 1993 ADR: 3,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...