EDTA tetrasodium salt hydrate 98

Supplier: BeanTown Chemical

Synonyms: Tetrasodium ethylenediaminetetraacetate hydrate, Kestranal® 4S hydrate, Ethylenedinitrilotetra-acetic acid tetrasodium salt hydrate, Sodium 2,2',2'',2'''-(ethane-1,2-diylbis(azanetriyl))tetraacetate xhydrate, Edetic acid tetrasodium salt hydrate, Ethylenediaminetetraacetic acid tetrasodium salt hydrate

142980-1KG 142980-250G 142980-5KG
BT142980-1KGEA 57 USD
BT142980-1KG BT142980-250G BT142980-5KG
EDTA tetrasodium salt hydrate 98
EDTA tetrasodium salt hydrate

CAS: 194491-31-1; EC No: 200-573-9; MDL No: MFCD00150025 Solid; Molecular Formula: C10H12N2Na4O8·xH2O ; MW: 380.17 (anhydrous) Melting Point: 300°


  • Solid

Formula: C₁₀H₁₂N₂O₈Na₄·xH₂O
MW: 380.17 g/mol (anhy)
Melting Pt: >300 °C
MDL Number: MFCD00150025
CAS Number: 13235-36-4
EINECS: 200-573-9

Order Now

Specification Test Results

Assay 98%

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR