Ethylene glycol ≥99.0% for spectrophotometry
Supplier: BeanTown Chemical
Synonyms:
Ethane-1,2-diol, 1,2-Dihydroxy ethane, 1,2-Ethanediol, Glycol
142950-1L
142950-4X1L
BT142950-1LEA
85.3
USD
BT142950-1L
BT142950-4X1L
Ethylene glycol ≥99.0% for spectrophotometry
Ethylene glycol
CAS: 107-21-1; EC No: 203-473-3; MDL No: MFCD00002885; RTECS: KW2975000
Liquid; Linear Formula: HOCH2CH2OH; MW: 62.07
Melting Point: -13°; Boiling Point: 196-198°; Flash point: 119°C (246°F)
Density (g/mL): 1.113; Refractive Index: 1.431
- RTECS: KW2975000
- Refractive index: 1.431
- Liquid
Formula:
HOCH₂CH₂OH MW: 62.07 g/mol Boiling Pt: 196…198 °C Melting Pt: –13 °C Density: 1.113 g/mL Flash Pt: 119 °C (246 °F) |
MDL Number:
MFCD00002885 CAS Number: 107-21-1 EINECS: 203-473-3 |
Specification Test Results
Assay | 99.0% min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...