2'-Fluoro-3'-(trifluoromethyl)acetophenone ≥97%
Supplier: BeanTown Chemical
Synonyms:
1-(2-Fluoro-3-(trifluoromethyl)phenyl)ethanone
142945-1G
142945-5G
BT142945-1GEA
86
USD
BT142945-1G
BT142945-5G
2'-Fluoro-3'-(trifluoromethyl)acetophenone ≥97%
2'-Fluoro-3'-(trifluoromethyl)acetophenone
CAS: 207853-63-2; MDL No: MFCD00061145
Liquid; Molecular Formula: C9H6F4O; MW: 206.14
Density (g/mL): 1.020
- Liquid
Formula:
C₉H₆F₄O MW: 206.14 g/mol Density: 1.020 g/mL |
MDL Number:
MFCD00061145 CAS Number: 207853-63-2 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...