Methyl-2-methylnicotinate ≥97%

Supplier: BeanTown Chemical

Synonyms: methyl 2-methylpyridine-3-carboxylate, Methyl 2-methylnicotinate

142920-25G 142920-5G
BT142920-25GEA 311 USD
BT142920-25G BT142920-5G
Methyl-2-methylnicotinate ≥97%
Methyl-2-methylnicotinate

CAS: 65719-09-7; MDL No: MFCD00051961 Liquid; Molecular Formula: C8H9NO2; MW: 151.16 Boiling Point: 102-104°/10 mmHg Refractive Index: 1.517


  • Refractive index: 1.517
  • Liquid

Formula: C₈H₉NO₂
MW: 151.16 g/mol
Boiling Pt: 102…104 °C/10 mmHg
MDL Number: MFCD00051961
CAS Number: 65719-09-7

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR