2-(1H-Imidazol-5-yl)acetic acid hydrochloride ≥97%
Supplier: BeanTown Chemical
Synonyms:
4-Imidazoleacetic acid hydrochloride, Imidazole-4-acetic acid monohydrochloride
142910-1G
142910-5G
BT142910-1GEA
121
USD
BT142910-1G
BT142910-5G
2-(1H-Imidazol-5-yl)acetic acid hydrochloride ≥97%
2-(1H-Imidazol-5-yl)acetic acid hydrochloride
CAS: 3251-69-2; EC No: 221-840-6; MDL No: MFCD00012698
Crystalline/Powder; Molecular Formula: C5H6N2O2·HCl; MW: 162.58
Melting Point: 218-222°
Hygroscopic
- Hygroscopic
- Crystalline/Powder
Formula:
C₅H₇ClN₂O₂ MW: 162.58 g/mol Melting Pt: 218…222 °C |
MDL Number:
MFCD00012698 CAS Number: 3251-69-2 EINECS: 221-840-6 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...