Lead ≥99.999% (trace metals basis), rods Ø 10 mm

Supplier: BeanTown Chemical

142895-1KG 142895-250G 142895-50G
BT142895-1KGEA 1812 USD
BT142895-1KG BT142895-250G BT142895-50G
Lead ≥99.999% (trace metals basis), rods Ø 10 mm
Lead

CAS: 7439-92-1; EC No: 231-100-4; MDL No: MFCD00134050; RTECS: OF7525000 Solid; Molecular Formula: Pb; MW: 207.20 Melting Point: 327.4°; Boiling Point: 1740° Density (g/mL): 11.36


  • RTECS: OF7525000
  • Solid

Formula: Pb
Boiling Pt: 1740 °C
Melting Pt: 327.4 °C
Density: 11.36 g/mL
MDL Number: MFCD00134050
CAS Number: 7439-92-1
EINECS: 231-100-4

Order Now

Specification Test Results

Assay 99.999%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR