1-Aminocyclopropanecarboxylic acid hydrochloride ≥97%
Supplier: BeanTown Chemical
Synonyms:
1-azanylcyclopropane-1-carboxylic acid hydrochloride, ACPC, 1-aminocyclopropane-1-carboxylic acid hydrochloride, Cyclopropanecarboxylic acid, 1-amino-, hydrochloride
142885-1G
142885-250MG
BT142885-1GEA
546
USD
BT142885-1G
BT142885-250MG
1-Aminocyclopropanecarboxylic acid hydrochloride ≥97%
1-Aminocyclopropanecarboxylic acid hydrochloride
CAS: 68781-13-5; MDL No: MFCD00012545
Solid; Molecular Formula: C4H7NO2·HCl ; MW: 137.56
Melting Point: 231° (decomposes)
Hygroscopic
- Hygroscopic
- Solid
Formula:
C₄H₈ClNO₂ MW: 137.57 g/mol Melting Pt: 231 °C (dec.) |
MDL Number:
MFCD00012545 CAS Number: 68781-13-5 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...