L(+)-Tartaric acid ≥99%

Supplier: BeanTown Chemical

Synonyms: L-Tartaric Acid, (2R,3R)-(+)-Tartaric acid, Tartaric Acid, (R,R)-(+)-2,3-Dihydroxysuccinic acid, (R,R)-(+)-Tartaric acid, (2R,3R)-2,3-Dihydroxysuccinic acid

142795-100G 142795-2.5KG 142795-500G
BT142795-100GEA 54.47 USD
BT142795-100G BT142795-2.5KG BT142795-500G
L(+)-Tartaric acid ≥99%
L(+)-Tartaric acid

CAS: 87-69-4; EC No: 201-766-0; MDL No: MFCD00064207; RTECS: WW7875000 Crystalline; Molecular Formula: C4H6O6; MW: 150.09 Melting Point: 170-172° Optical Rotation: [α]20/D +13.5±0.5°, c = 10% in water


  • RTECS: WW7875000
  • Optical rotation: [α]20/D +13.5±0.5°, c = 10% in water
  • Crystalline

Formula: C₄H₆O₆
Melting Pt: 170…172 °C
MDL Number: MFCD00064207
CAS Number: 87-69-4
EINECS: 201-766-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4e859e52081482a351d52322717614aad450742&vid=879c292903ddee7a59d5d6b2d969c4c8fc6eac75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR