(-)-Dibenzoyl-L-tartaric acid monohydrate ≥98%

Supplier: BeanTown Chemical

Synonyms: (2R,3R)-(-)-Di-O-benzoyl-tartaric acid monohydrate

142750-100G 142750-500G
BT142750-100GEA 53 USD
BT142750-100G BT142750-500G
(-)-Dibenzoyl-L-tartaric acid monohydrate ≥98%
(-)-Dibenzoyl-L-tartaric acid monohydrate

CAS: 62708-56-9; EC No: 220-374-0; MDL No: MFCD00149119; RTECS: WW8070000 Solid; Molecular Formula: C18H14O8·H2O; MW: 376.31 Melting Point: 88-89° Optical Rotation: [α]20/D −110°, c = 5% in methanol


  • RTECS: WW8070000
  • Optical rotation: [α]20/D −110°, c = 5% in methanol
  • Solid

Formula: C₁₈H₁₄O₈·1H₂O
Melting Pt: 88…89 °C
MDL Number: MFCD00149119
CAS Number: 62708-56-9
EINECS: 220-374-0

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR