Lithium tetrachloropalladate(II)
Supplier: BeanTown Chemical
142745-1G
142745-5G
BT142745-1GEA
133
USD
BT142745-1G
BT142745-5G
Lithium tetrachloropalladate(II)
Lithium tetrachloropalladate(II)
CAS: 15525-45-8; EC No: 239-567-6; MDL No: MFCD00011082
Crystalline; Linear Formula: Li2PdCl4; MW: 262.09
Moisture Sensitive
- Moisture Sensitive
- Crystalline
Formula:
Cl₄Li₂Pd MW: 262.1 g/mol |
MDL Number:
MFCD00011082 CAS Number: 15525-45-8 EINECS: 239-567-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...