Dibenzo-18-crown-6-(dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, tin(II) ionophore I) ≥98%
Supplier: BeanTown Chemical
Synonyms:
Dibenzo-18-crown-6
142735-25G
142735-5G
BT142735-25GEA
76
USD
BT142735-25G
BT142735-5G
Dibenzo-18-crown-6-(dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, tin(II) ionophore I) ≥98%
Dibenzo-18-crown-6-(dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, tin(II) ionophore I)
CAS: 14187-32-7; EC No: 238-041-3; MDL No: MFCD00005098; RTECS: HP5386000
Powder; Molecular Formula: C20H24O6; MW: 360.41
Melting Point: 162-164°
- RTECS: HP5386000
- Powder
Formula:
C₂₀H₂₄O₆ MW: 360.41 g/mol Melting Pt: 162…164 °C |
MDL Number:
MFCD00005098 CAS Number: 14187-32-7 EINECS: 238-041-3 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...