Silver ≥99.9% (trace metals basis), foil, annealed, Thickness 1.0 mm
Supplier: BeanTown Chemical
142695-100X100
142695-25X25MM
142695-50X50MM
BT142695-100X1EA
651.25
USD
BT142695-100X1
BT142695-25X25
BT142695-50X50
Silver ≥99.9% (trace metals basis), foil, annealed, Thickness 1.0 mm
Silver
CAS: 7440-22-4; EC No: 231-131-3; MDL No: MFCD00003397
Solid; Molecular Formula: Ag; MW: 107.87
Melting Point: 960°; Boiling Point: 2212°
Density (g/mL): 10.49
- Solid
Formula:
Ag Boiling Pt: 2212 °C Melting Pt: 960 °C Density: 10.49 g/mL |
MDL Number:
MFCD00003397 CAS Number: 7440-22-4 EINECS: 231-131-3 |
Specification Test Results
Assay | 99.9% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...