4-Chlororesorcinol ≥98%
Supplier: BeanTown Chemical
142655-100G
142655-25G
142655-500G
BT142655-100GEA
40
USD
BT142655-100G
BT142655-25G
BT142655-500G
4-Chlororesorcinol ≥98%
4-Chlororesorcinol
CAS: 95-88-5; EC No: 202-462-0; MDL No: MFCD00002273; RTECS: VH0450000
Powder; Molecular Formula: C6H5ClO2; MW: 144.56
Melting Point: 106-108°; Boiling Point: 147°/18 mmHg
- RTECS: VH0450000
- Powder
Formula:
C₆H₅ClO₂ Boiling Pt: 147 °C/18 mmHg Melting Pt: 106…108 °C |
MDL Number:
MFCD00002273 CAS Number: 95-88-5 EINECS: 202-462-0 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...