12-Tricosanone ≥97%
Supplier: BeanTown Chemical
142610-100G
142610-25G
BT142610-100GEA
254
USD
BT142610-100G
BT142610-25G
12-Tricosanone ≥97%
12-Tricosanone
CAS: 540-09-0; EC No: 208-735-0; MDL No: MFCD00026562; RTECS: YD0805000
Powder; Linear Formula: [CH3(CH2)10]2CO; Molecular Formula: C23H46O; MW: 338.62
- RTECS: YD0805000
- Powder
Formula:
C₂₃H₄₆O |
MDL Number:
MFCD00026562 CAS Number: 540-09-0 EINECS: 208-735-0 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...