Iron(III) oxide ≥99% (trace metals basis)

Supplier: BeanTown Chemical

142605-10G 142605-50G
BT142605-10GEA 232 USD
BT142605-10G BT142605-50G
Iron(III) oxide ≥99% (trace metals basis)
Iron(III) oxide

CAS: 1309-37-1; EC No: 215-168-2; MDL No: MFCD00011008 8-10nm APS Powder; Molecular Formula: Fe2O3 ; MW: 159.69


  • 8-10nm APS Powder

Formula: Fe₂O₃
MDL Number: MFCD00011008
CAS Number: 1309-37-1
EINECS: 215-168-2

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR