Abietic acid ≥90%, Technical Grade

Supplier: BeanTown Chemical

142600-1G 142600-250MG
BT142600-1GEA 272.52 USD
BT142600-1G BT142600-250MG
Abietic acid ≥90%, Technical Grade
Abietic acid

CAS: 514-10-3; EC No: 208-178-3; MDL No: MFCD03423567; RTECS: TP8580000 Crystalline; Molecular Formula: C20H30O2; MW: 302.45 Melting Point: 139-142° Air Sensitive


  • RTECS: TP8580000
  • Air Sensitive
  • Crystalline

Formula: C₂₀H₃₀O₂
Melting Pt: 139…142 °C
MDL Number: MFCD03423567
CAS Number: 514-10-3
EINECS: 208-178-3

Order Now

Specification Test Results

Assay 90%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR