4-Amino-3-hydroxy-1-naphthalenesulfonic acid ≥90% ACS
Supplier: BeanTown Chemical
Synonyms:
1-Amino-2-naphthol-4-sulphonic acid, 4-Amino-3-hydroxy-α-naphthalenesulphonic acid, 4-Amino-3-hydroxynaphthalene-1-sulphonic acid, 1-Amino-2-hydroxy-4-naphthalenesulphonic acid
142590-100G
142590-25G
142590-5G
BT142590-100GEA
162
USD
BT142590-100G
BT142590-25G
BT142590-5G
4-Amino-3-hydroxy-1-naphthalenesulfonic acid ≥90% ACS
4-Amino-3-hydroxy-1-naphthalenesulphonic acid
CAS: 116-63-2; EC No: 204-147-3; MDL No: MFCD00004019; RTECS: QK1292000
Powder; Molecular Formula: C10H9NO4S; MW: 239.25
Melting Point: 290° (decomposes)
- RTECS: QK1292000
- Powder
Formula:
C₁₀H₉NO₄S MW: 239.25 g/mol Melting Pt: 290 °C (dec.) |
MDL Number:
MFCD00004019 CAS Number: 116-63-2 EINECS: 204-147-3 |
Specification Test Results
Assay | 98.0% min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...