Diphenyltin dichloride ≥95%

Supplier: BeanTown Chemical

Synonyms: Dichlorodiphenylstannane

142585-25G 142585-5G
BT142585-25GEA 93 USD
BT142585-25G BT142585-5G
Diphenyltin dichloride ≥95%
Diphenyltin dichloride

CAS: 1135-99-5; EC No: 214-496-3; MDL No: MFCD00000516; RTECS: WH7253000 Crystalline; Linear Formula: (C6H5)2SnCl2; MW: 343.81 Melting Point: 41-43°; Boiling Point: 333-337°; Flash point: 110°C (230°F) Moisture Sensitive


  • RTECS: WH7253000
  • Moisture Sensitive
  • Crystalline

Formula: C₁₂H₁₀Cl₂Sn
MW: 343.83 g/mol
Boiling Pt: 333…337 °C
Melting Pt: 41…43 °C
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00000516
CAS Number: 1135-99-5
EINECS: 214-496-3
UN: 3146
ADR: 6.1,III

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR