1-(2-Methoxyphenoxy)-2,3-epoxypropane

Supplier: BeanTown Chemical

142565-25G 142565-5G
BT142565-25GEA 352 USD
BT142565-25G BT142565-5G
1-(2-Methoxyphenoxy)-2,3-epoxypropane
1-(2-Methoxyphenoxy)-2,3-epoxypropane

CAS: 2210-74-4; EC No: 218-644-8; MDL No: MFCD00190158; RTECS: TZ3550000 Solid; Molecular Formula: C10H12O3 ; MW: 180.20


  • RTECS: TZ3550000
  • Solid

Formula: C₁₀H₁₂O₃
MDL Number: MFCD00190158
CAS Number: 2210-74-4
EINECS: 218-644-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR