Hydroxypropyl methylcellulose (Hypromellose)
Supplier: BeanTown Chemical
Synonyms:
Methocel K100, Hydroxypropyl methyl cellulose
142550-1KG
142550-250G
142550-50G
BT142550-1KGEA
273.32
USD
BT142550-1KG
BT142550-250G
BT142550-50G
Hydroxypropyl methylcellulose (Hypromellose)
Hydroxypropylmethylcellulose
CAS: 9004-65-3; MDL No: MFCD00131360
Powder
- Powder
Formula:
C₃H₈O₂·xCH₄O |
MDL Number:
MFCD00131360 CAS Number: 9004-65-3 Merck Index: 14,4842 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...