Aluminium nitride

Supplier: BeanTown Chemical

Synonyms: Aluminum nitride

142545-250G 142545-50G
BT142545-250GEA 152 USD
BT142545-250G BT142545-50G
Aluminium nitride
Aluminium nitride

CAS: 24304-00-5; EC No: 246-140-8; MDL No: MFCD00003429; RTECS: BD1055000 -325 Mesh Powder, typically 4 Micron APS K19; Molecular Formula: AlN; MW: 40.99 Melting Point: 2200° Density (g/mL): 3.26 Moisture Sensitive


  • RTECS: BD1055000
  • Moisture Sensitive
  • -325 Mesh Powder, typically <4 Micron APS K19

Formula: AlN
MW: 40.99 g/mol
Melting Pt: >2200 °C
Density: 3.26 g/mL
MDL Number: MFCD00003429
CAS Number: 24304-00-5
EINECS: 246-140-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR