1,4-Benzodioxane ≥98%

Supplier: BeanTown Chemical

Synonyms: 2,3-Dihydrobenzo[b][1,4]dioxine, 2,3-Dihydro-1,4-benzodioxin, Benzo-1,4-dioxane, 1,4-Benzodioxan

142535-10G 142535-250G 142535-50G
BT142535-10GEA 26 USD
BT142535-10G BT142535-250G BT142535-50G
1,4-Benzodioxane ≥98%
1,4-Benzodioxane

CAS: 493-09-4; EC No: 207-775-6; MDL No: MFCD00006821; RTECS: DF4728000 Liquid; Molecular Formula: C8H8O2; MW: 136.15 Boiling Point: 216°; Flash point: 87°C (188°F) Density (g/mL): 1.17; Refractive Index: 1.549


  • RTECS: DF4728000
  • Refractive index: 1.549
  • Liquid

Formula: C₈H₈O₂
MW: 136.15 g/mol
Boiling Pt: 216 °C
Density: 1.17 g/mL
Flash Pt: 87 °C (188 °F)
MDL Number: MFCD00006821
CAS Number: 493-09-4
EINECS: 207-775-6

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR