Ethyl-2-phenylacrylate ≥94%
Supplier: BeanTown Chemical
Synonyms:
Ethyl 2-phenylacrylate
142515-1G
142515-25G
142515-5G
BT142515-1GEA
88
USD
BT142515-1G
BT142515-25G
BT142515-5G
Ethyl-2-phenylacrylate ≥94%
Ethyl-2-phenylacrylate
CAS: 22286-82-4; EC No: 244-892-1; MDL No: MFCD00026862
Liquid; Molecular Formula: C11H12O2; MW: 176.22
Boiling Point: 123-125°/16 mmHg
Light Sensitive
- Light Sensitive
- Liquid
Formula:
C₁₁H₁₂O₂ MW: 176.215 g/mol Boiling Pt: 123…125 °C/16 mmHg |
MDL Number:
MFCD00026862 CAS Number: 22286-82-4 EINECS: 244-892-1 |
Specification Test Results
Assay | 94% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...