L-Phenylalanine ≥99%
Supplier: BeanTown Chemical
Synonyms:
H-L-Phe-OH, Phenylalanine, H-Phe-OH, L-α-Phenylalanine, (S)-(-)-Phenylalanine, 3-Phenyl-L-alanine, 3-Phenyl-L(-)-alanine
142490-100G
142490-25G
BT142490-100GEA
66.82
USD
BT142490-100G
BT142490-25G
L-Phenylalanine ≥99%
L-Phenylalanine
CAS: 63-91-2; EC No: 200-568-1; MDL No: MFCD00064227; RTECS: AY7535000
Crystalline; Molecular Formula: C9H11NO2; MW: 165.19
Melting Point: 270-275° (decomposes)
Optical Rotation: [α]20/D -34±0.5°, c = 2% in water
- RTECS: AY7535000
- Optical rotation: [α]20/D -34±0.5°, c = 2% in water
- Crystalline
Formula:
C₉H₁₁NO₂ MW: 165.19 g/mol Melting Pt: 270…275 °C (dec.) |
MDL Number:
MFCD00064227 CAS Number: 63-91-2 EINECS: 200-568-1 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...