7-Chloro-4-quinazolone ≥97%

Supplier: BeanTown Chemical

Synonyms: 7-Chloro-4-hydroxyquinazoline

142465-1G 142465-5G
BT142465-1GEA 55 USD
BT142465-1G BT142465-5G
7-Chloro-4-quinazolone ≥97%
7-Chloro-4-quinazolone

CAS: 31374-18-2 Solid; Molecular Formula: C8H5ClN2O; MW: 180.59


  • Solid

Formula: C₈H₅ClN₂O
MW: 180.5900 g/mol
MDL Number: MFCD01570890
CAS Number: 31374-18-2

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR